Learn More
Ytterbium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Cl3H2OYb, CAS Number-19423-87-1, acmc-1bo6s, ytterbium 3+ chloride-water 1/3/1, ytterbium 3+ ion hydrate trichloride, ytterbium iii chloride hydrate, 50g, 99.99% (REO), White, 2.58, 233-800-5, 279.40 (Anhydrous), H315-H319-H335
£78.89 - £287.43
Chemical Identifiers
CAS | 19423-87-1 |
---|---|
Molecular Formula | Cl3Yb |
Molecular Weight (g/mol) | 279.40 |
MDL Number | MFCD00149885 |
InChI Key | CKLHRQNQYIJFFX-UHFFFAOYSA-K |
Synonym | ytterbium iii chloride hydrate, acmc-1bo6s, ytterbium 3+ ion hydrate trichloride, ytterbium 3+ chloride-water 1/3/1 |
PubChem CID | 71317291 |
IUPAC Name | ytterbium(3+);trichloride;hydrate |
SMILES | [Cl-].[Cl-].[Cl-].[Yb+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356228
|
Thermo Scientific Alfa Aesar
011194.09 |
10 g |
£78.89
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346228
|
Thermo Scientific Alfa Aesar
011194.18 |
50 g |
£287.43
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ytterbium(III) chloride hydrate is used as a catalyst in organic chemical synthesis. It is also used to track digestion in animals. The catalytic nature of this compound also has an application in DNA microarrays.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
19423-87-1 | |
279.40 | |
CKLHRQNQYIJFFX-UHFFFAOYSA-K | |
71317291 | |
[Cl-].[Cl-].[Cl-].[Yb+3] |
Cl3Yb | |
MFCD00149885 | |
ytterbium iii chloride hydrate, acmc-1bo6s, ytterbium 3+ ion hydrate trichloride, ytterbium 3+ chloride-water 1/3/1Show More | |
ytterbium(3+);trichloride;hydrate |
Specifications
150°C to 155°C | |
White | |
99.99% (REO) | |
MFCD00149885 | |
Soluble in water. | |
[Cl-].[Cl-].[Cl-].[Yb+3] | |
279.40 | |
279.40 (Anhydrous) | |
Hygroscopic | |
Ytterbium(III) chloride hydrate |
19423-87-1 | |
Crystalline Aggregates | |
Cl3Yb | |
ytterbium iii chloride hydrate, acmc-1bo6s, ytterbium 3+ ion hydrate trichloride, ytterbium 3+ chloride-water 1/3/1 | |
CKLHRQNQYIJFFX-UHFFFAOYSA-K | |
ytterbium(3+);trichloride;hydrate | |
71317291 | |
Odorless | |
2.58 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 233-800-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.