missing translation for 'onlineSavingsMsg'
Learn More
Ytterbium powder, -200 mesh, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 15482567

Ytterbium powder, -200 mesh, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 15482567
2 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 15482567

Brand: Thermo Scientific Alfa Aesar 045920.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-64-4
Molecular Formula Yb
Molecular Weight (g/mol) 173.05
MDL Number MFCD00011286
InChI Key NAWDYIZEMPQZHO-UHFFFAOYSA-N
Synonym unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
PubChem CID 23992
ChEBI CHEBI:33381
IUPAC Name ytterbium
SMILES [Yb]

Specifications

Color Silver
UN Number UN3089
Quantity 2 g
Percent Purity 99.9%
Mesh Size −200 Mesh
Odor Odorless
Sensitivity Air and moisture sensitive
Form Powder
Assay (REO)
Chemical Name or Material Ytterbium powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR03K004 Certificate TypeCertificate of Analysis Date08/05/2024 Catalog Number045920.09,045920.18,045920.04
Lot NumberU24J010 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number045920.09,045920.18,045920.04
Lot NumberM25J011 Certificate TypeCertificate of Analysis Date30/01/2023 Catalog Number045920.09,045920.18,045920.04
Lot NumberS21I038 Certificate TypeCertificate of Analysis Date11/07/2022 Catalog Number045920.09,045920.18,045920.04
Lot NumberN25G028 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045920.09,045920.18,045920.04
compliance-icons
Product Identifier
  • Ytterbium powder
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H228-Flammable solid.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Ytterbium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ytterbium powder, -200 mesh, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.