missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) chloride hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

£68.60 - £218.00

Chemical Identifiers

CAS 10025-94-2
Molecular Formula Cl36H2O
Molecular Weight (g/mol) 303.35
MDL Number MFCD00149941
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10420671
View Documents Promotion Details
Thermo Scientific Acros
199180500
50 g Glass bottle
£68.60
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10216640
View Documents Promotion Details
Thermo Scientific Acros
199182500
250 g Glass bottle
£218.00
250g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
Specifications

Chemical Identifiers

10025-94-2
Cl36H2O
303.35
MFCD00149941

Specifications

100.0°C
White
99.90%
YCl3·6H2O
14, 10107
303.35
99.9%
Yttrium(III) chloride hexahydrate, 99.9%
10025-94-2
Adhering Crystalline Powder or Crystals
Cl36H2O
MFCD00149941
Solubility in water: soluble. Other solubilities: soluble in alcohols
303.35
Trace Metal Basis
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 233-801-0

missing translation for 'rtecsNumber' : ZG3150000

missing translation for 'tsca' : TSCA

Product Identifier
  • Yttrium(III) chloride hexahydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only