Learn More
Yttrium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Description
It acts as a precursor for the preparation of yttrium oxide nano particles and for nanoscale coatings of carbon composites. It is an effective yttrium source for the preparation of yttrium-based surfactant mesophases which are used as adsorbing agents and in optically functional materials. It is used in optics, ceramics, glass, electronics, and superconducting materials. It has been reported as a reusable catalyst in Bignelli reaction for the synthesis of heterocycles such as pyrimidin-2-ones. It is a powerful catalyst for the synthesis of some supramolecules such as calix[4]resorcinarenes, 1,8-dioxooctahydroxanthenes, 2-amino-4H-chromenes, and several other organic condensation products. Silica-supported yttrium nitrate hydrate acts as a heterogeneous catalyst for the preparation of 2-Amino-4H-Chromenes in water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Crystalline Aggregates |
Quantity | 5 x 100 g |
UN Number | UN1477 |
Merck Index | 14,10107 |
Solubility Information | Soluble in water. |
IUPAC Name | yttrium(3+);trinitrate;hydrate |
Formula Weight | 274.92 (Anhydrous) |
Percent Purity | 99.99% |
Grade | Reacton™ |
Sensitivity | Hygroscopic |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
S05K067 | Certificate of Analysis | 25/06/2024 | 011187.22,011187.06,011187.14,011187.59 |
Y09G053 | Certificate of Analysis | 11/06/2024 | 011187.22,011187.06,011187.14,011187.59 |
X11J010 | Certificate of Analysis | 24/10/2023 | 011187.22,011187.06,011187.14,011187.59 |
61200296 | Certificate of Analysis | 23/11/2022 | 011187.22,011187.06,011187.14,011187.59 |
61200296 | Certificate of Analysis | 22/11/2022 | 011187.22,011187.06,011187.14,011187.59 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.