Learn More
Yttrium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Description
Yttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is presurcor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 2410°C |
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Linear Formula | Y2O3 |
Merck Index | 14,10107 |
Solubility Information | Insoluble in water. Soluble in alcohol and dilute acids |
Formula Weight | 225.81 |
Percent Purity | 99.99% |
Odor | Odorless |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N21L023 | Certificate of Analysis | 24/03/2025 | 011181.A1,011181.22,011181.14,011181.36 |
Z24K049 | Certificate of Analysis | 26/12/2024 | 011181.22,011181.A1,011181.14,011181.36 |
T25K049 | Certificate of Analysis | 20/08/2024 | 011181.22,011181.A1,011181.14,011181.36 |
P18K033 | Certificate of Analysis | 10/05/2024 | 011181.22,011181.A1,011181.14,011181.36 |
W28J018 | Certificate of Analysis | 05/10/2023 | 011181.22,011181.A1,011181.14,011181.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.