missing translation for 'onlineSavingsMsg'
Learn More
Yttrium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals Product Code.: 11343858

Yttrium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

Product Code. 11343858
100 mm
Click to view available options
Quantity:
25 mm
50 mm
100 mm
This item is not returnable. View return policy

Product Code. 11343858

Brand: Thermo Scientific Alfa Aesar 040300.DM

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-65-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
Synonym yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]

Specifications

Quantity 100 mm
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.57g/mm
Form Cast Rod
Assay (metals basis excluding Ta)
Chemical Name or Material Yttrium rod, 12.7mm (0.5 in.) dia.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA24Y012 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040300.DJ,040300.DH,040300.DM
Lot Number61101200 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040300.DJ,040300.DH,040300.DM
Lot NumberH31L16 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040300.DJ,040300.DH,040300.DM
Lot NumberJ18Q083 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040300.DJ,040300.DH,040300.DM
Lot NumberF08L25 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040300.DJ,040300.DH,040300.DM

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.