Learn More
ZC3H12A Mouse, anti-Human, Clone: 2F6B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68616-1-IG-150UL
Additional Details : Weight : 0.00100kg
Description
The ZC3H12A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZC3H12A. This antibody recognizes human antigen. The ZC3H12A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
ZC3H12A | |
2F6B2 | |
dJ423B22.1, FLJ23231, MCP induced protein 1, MCPIP, MCPIP1, Ribonuclease ZC3H12A, RP3 423B22.1, ZC3H12A | |
150 μL | |
80149 | |
Store at -20°C. Stable for one year after shipment. | |
Liquid |
Western Blot, ELISA | |
1000 μg/mL | |
Mouse | |
Primary | |
Human | |
150 kDa | |
IgG1 |