missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H12A Mouse, anti-Human, Clone: 2F6B2, Proteintech
Mouse Monoclonal Antibody
£155.00 - £404.20
Specifications
Antigen | ZC3H12A |
---|---|
Clone | 2F6B2 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18079435
|
Proteintech
68616-1-IG-20UL |
20 μL |
£155.00
20µL |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18019835
|
Proteintech
68616-1-IG-150UL |
150 μL |
£404.20
150µL |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The ZC3H12A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZC3H12A. This antibody recognizes human antigen. The ZC3H12A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
ZC3H12A | |
1000 μg/mL | |
Liquid | |
Human | |
dJ423B22.1, FLJ23231, MCP induced protein 1, MCPIP, MCPIP1, Ribonuclease ZC3H12A, RP3 423B22.1, ZC3H12A | |
Primary |
2F6B2 | |
Western Blot, ELISA | |
Mouse | |
80149 | |
IgG1 | |
Store at -20°C. Stable for one year after shipment. |