missing translation for 'onlineSavingsMsg'
Learn More

ZVG™ ZetGigant™ Small Toilet Paper Rolls

ZetGigant™ Tissue Toilet Paper in small rolls provides high quality and comfort. With perforated tissue paper and small rolls, it offers easy handling.

Brand:  ZVG™ 19010-04

 View more versions of this product

Product Code. 17873545

  • £77.75 / 48 rolls
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

ZVG™ offers a selected range of voluminous, absorbent, and sturdy toilet papers. The high-yield zetGigant™ roll , which sets new economic standards, proves its true size: seldom refilling, reduced maintenance times and constant operational readiness.

  • Small rolls
  • Perforated
  • With or without floral design
  • Each roll contains 250 sheets
Specifications

Specifications

Toilet Paper
11 cm
9.5 cm
White
4.3 in.
3.7 in.
Recycled Fiber
2-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.