Learn More
ZVG™ zetRoll™ Hand Towel Rolls
The zetRoll™ Hand Towel Roll provides high absorbency and allows for hygienic hand drying. With its dot-to-dot embossing, it is particularly convenient to use.
Brand: ZVG™ 16463-06
Description
The high-quality zetRoll™ disposable paper towel rolls are ideal for gastronomy, schools, care facilities, kitchens, and essential at gas stations for thorough removal of dirt, oil, and fuel residues. They provide a quick and convenient way to dry hands or quickly wipe surfaces or objects in the vicinity. All 2-ply and 3-ply paper towel rolls have point-to-point embossing.
- White
- High absorbency
- Hygienic hand drying
- With point-to-point embossing
Point-to-point embossing: In a novel manufacturing process, the individual layers of the paper towel are bonded together at multiple points across the entire surface. This creates a unified structure that is highly tear-resistant. This process combines the advantages of a thick wipe (stability) with those of a multi-layered towel (high absorbency).
Specifications
Towel | |
153 m | |
20.3 cm | |
White | |
2-ply |
13.4 in. | |
8 in. | |
Cellulose | |
Single-use |