missing translation for 'onlineSavingsMsg'
Learn More

Zinc 2,4-pentanedionate monohydrate, Thermo Scientific Chemicals

£25.83 - £74.90

Chemical Identifiers

CAS 14363-15-6
Molecular Formula C10H16O5Zn
Molecular Weight (g/mol) 281.61
MDL Number MFCD00000035
InChI Key BNHYOPPPENBGNL-SUKNRPLKSA-L
Synonym zinc 2,4-pentanedionate monohydrate
PubChem CID 122130627
IUPAC Name zinc(2+) (2Z)-pent-2-ene-2,4-bis(olate) (2Z)-4-oxopent-2-en-2-olate hydrate
SMILES O.[Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=[O-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11381859
View Documents
Thermo Scientific Alfa Aesar
012527.18
50 g
£25.83
50g
Please to purchase this item. Need a web account? Register with us today!
11391859
View Documents
Thermo Scientific Alfa Aesar
012527.30
250 g
£74.90
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zinc Metal acetylacetonates are used as catalysts for polymerization of olefins and transesterification. It is used as a a stabilizer PVC.They are used as solvents, lubricant additives, paint drier, and pesticides. They are used in glass coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14363-15-6
281.61
BNHYOPPPENBGNL-SUKNRPLKSA-L
122130627
O.[Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=[O-]
C10H16O5Zn
MFCD00000035
zinc 2,4-pentanedionate monohydrate
zinc(2+) (2Z)-pent-2-ene-2,4-bis(olate) (2Z)-4-oxopent-2-en-2-olate hydrate

Specifications

14363-15-6
1.6 g/mL
MFCD00000035
Soluble in alcohol; Slightly soluble in water
O.[Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=[O-]
281.61
281.60 (263.59 Anhydrous)
Zinc 2,4-pentanedionate monohydrate
136°C to 138°C (decomposition)
C10H16O5Zn
zinc 2,4-pentanedionate monohydrate
BNHYOPPPENBGNL-SUKNRPLKSA-L
zinc(2+) (2Z)-pent-2-ene-2,4-bis(olate) (2Z)-4-oxopent-2-en-2-olate hydrate
122130627
Crystalline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 237-860-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only