missing translation for 'onlineSavingsMsg'
Learn More

Zinc bromide, anhydrous, 98%, Thermo Scientific Chemicals

Zinc bromide, 98%, Br2Zn, CAS Number-7699-45-8, acmc-209p7h, zinc bromide, anhydrous 100g, zincbromide, zinc bromide hydrate, puratronic, ksc378m6n, zinc 2+ ion dibromide, zinc ii bromide, 500g, EM>9,, 650 deg.C, 4.201

£30.15 - £92.65

Chemical Identifiers

CAS 7699-45-8
Molecular Formula Br2Zn
Molecular Weight (g/mol) 225.188
MDL Number MFCD00011294
InChI Key VNDYJBBGRKZCSX-UHFFFAOYSA-L
Synonym zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100gShow MoreShow Less
PubChem CID 10421208
IUPAC Name zinc;dibromide
SMILES [Zn+2].[Br-].[Br-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11470697
SDS View Documents
Thermo Scientific Alfa Aesar
B22510.22
100 g
£30.15
100g
Please to purchase this item. Need a web account? Register with us today!
11480697
SDS View Documents
Thermo Scientific Alfa Aesar
B22510.36
500 g
£92.65
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zinc bromide solution can be used as a transparent shield against radiation. It is used as an optimal catalyst for stereospecific and regioselective reaction of silacyclopropanes with carbonyl compounds. It finds an application as Lewis acid in organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc bromide solution can be used as a transparent shield against radiation. It is used as an optimal catalyst for stereospecific and regioselective reaction of silacyclopropanes with carbonyl compounds. It finds an application as Lewis acid in organic reactions.

Solubility
Soluble in alcohol, ether, acetone and tetrahydrofuran.

Notes
Hygroscopic.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zinc bromide, anhydrous, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.