missing translation for 'onlineSavingsMsg'
Learn More
Zinc bromide, anhydrous, 98%, Thermo Scientific Chemicals Product Code.: 11470697

Zinc bromide, anhydrous, 98%, Thermo Scientific Chemicals

Product Code. 11470697
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11470697

Brand: Thermo Scientific Alfa Aesar B22510.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zinc bromide, 98%, Br2Zn, CAS Number-7699-45-8, acmc-209p7h, zinc bromide, anhydrous 100g, zincbromide, zinc bromide hydrate, puratronic, ksc378m6n, zinc 2+ ion dibromide, zinc ii bromide, 500g, EM>9,, 650 deg.C, 4.201

Zinc bromide solution can be used as a transparent shield against radiation. It is used as an optimal catalyst for stereospecific and regioselective reaction of silacyclopropanes with carbonyl compounds. It finds an application as Lewis acid in organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc bromide solution can be used as a transparent shield against radiation. It is used as an optimal catalyst for stereospecific and regioselective reaction of silacyclopropanes with carbonyl compounds. It finds an application as Lewis acid in organic reactions.

Solubility
Soluble in alcohol, ether, acetone and tetrahydrofuran.

Notes
Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7699-45-8
Molecular Formula Br2Zn
Molecular Weight (g/mol) 225.188
MDL Number MFCD00011294
InChI Key VNDYJBBGRKZCSX-UHFFFAOYSA-L
Synonym zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100gShow MoreShow Less
PubChem CID 10421208
IUPAC Name zinc;dibromide
SMILES [Zn+2].[Br-].[Br-]

Specifications

Boiling Point 650°C
Melting Point 394°C
Quantity 100 g
UN Number UN3260
Beilstein EM>9,
Merck Index 14,10129
Solubility Information Soluble in alcohol,ether,acetone and tetrahydrofuran.
Formula Weight 225.19
Percent Purity 98%
Refractive Index 1.5452
Sensitivity Hygroscopic
Density 4.201
Chemical Name or Material Zinc bromide, anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252590 Certificate TypeCertificate of Analysis Date12/11/2024 Catalog NumberB22510.22,B22510.36
Lot Number10246755 Certificate TypeCertificate of Analysis Date25/05/2023 Catalog NumberB22510.22
Lot Number10242485 Certificate TypeCertificate of Analysis Date28/07/2022 Catalog NumberB22510.22,B22510.36
Lot Number10201976 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22510.22,B22510.36
Lot Number10156296 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22510.22,B22510.36
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc bromide, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Zinc bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zinc bromide, anhydrous, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.