missing translation for 'onlineSavingsMsg'
Learn More
Zinc fluoride, anhydrous, 99% min, Thermo Scientific Chemicals Product Code.: 11387237

Zinc fluoride, anhydrous, 99% min, Thermo Scientific Chemicals

Product Code. 11387237
50 g
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11387237

Brand: Thermo Scientific Alfa Aesar 011541.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is widely used in fluorination of organic compounds, as additive of electrolytic aluminum and also in electroplating baths. It is used in preserving food, manufacture of flux, oxygen-sensitive applications such as metal production, glasses, enamels, fabrics and plastics. It is also used in oil refining, manufacture of pharmaceuticals, manufacture phosphors for fluorescent electric lights, and floor coverings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-49-5
Molecular Formula F2Zn
Molecular Weight (g/mol) 103.377
MDL Number MFCD00011298
InChI Key BHHYHSUAOQUXJK-UHFFFAOYSA-L
Synonym zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride
PubChem CID 10129912
IUPAC Name zinc;difluoride
SMILES [F-].[F-].[Zn+2]

Specifications

Boiling Point 1500°C
Melting Point 872°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99% min. (anhydrous)
Solubility Information Sparingly soluble in hydrochloric acid,nitric acid,and ammonium hydroxide.
Formula Weight 103.37
Odor Odorless
Density 4.95
Chemical Name or Material Zinc fluoride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU15K033 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog Number011541.18,011541.A1,011541.30
Lot NumberW09K036 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog Number011541.18,011541.A1,011541.30
Lot NumberP15K056 Certificate TypeCertificate of Analysis Date21/03/2024 Catalog Number011541.18,011541.A1,011541.30
Lot NumberZ12J058 Certificate TypeCertificate of Analysis Date22/01/2024 Catalog Number011541.18,011541.A1,011541.30
Lot NumberS09J021 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number011541.18,011541.A1,011541.30
compliance-icons compliance-icons
Product Identifier
  • Zinc fluoride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Zinc fluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zinc fluoride, anhydrous, 99% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.