Learn More
Zinc nitride, 99% (metals basis), Thermo Scientific Chemicals
£63.30 - £935.00
Chemical Identifiers
CAS | 1313-49-1 |
---|---|
Molecular Formula | N2Zn3 |
Molecular Weight (g/mol) | 224.15 |
MDL Number | MFCD00061327 |
InChI Key | AKJVMGQSGCSQBU-UHFFFAOYSA-N |
Synonym | zinc nitride, zinc nitride zn3n2 |
PubChem CID | 12130759 |
SMILES | [Zn++].[N-]=[Zn].[N-]=[Zn] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11329219
|
Thermo Scientific Alfa Aesar
088212.04 |
2 g |
£63.30
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11319219
|
Thermo Scientific Alfa Aesar
088212.09 |
10 g |
£272.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15412987
|
Thermo Scientific Alfa Aesar
088212.18 |
50 g | |||||||
Description
Zinc nitride is useful in optical and electronic devices. It is doped with oxygen to prepare a potential nitride-based transparent conductor, which is used in semiconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1313-49-1 | |
224.15 | |
AKJVMGQSGCSQBU-UHFFFAOYSA-N | |
12130759 |
N2Zn3 | |
MFCD00061327 | |
zinc nitride, zinc nitride zn3n2 | |
[Zn++].[N-]=[Zn].[N-]=[Zn] |
Specifications
1313-49-1 | |
N2Zn3 | |
zinc nitride, zinc nitride zn3n2 | |
AKJVMGQSGCSQBU-UHFFFAOYSA-N | |
zinc;azanidylidenezinc | |
12130759 | |
99% | |
(metals basis) |
Powder | |
MFCD00061327 | |
Insoluble in water. Soluble in hydrochloric acid. | |
[Zn++].[N-]=[Zn].[N-]=[Zn] | |
224.15 | |
224.12 | |
6.22 g/mL | |
Zinc nitride |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 215-207-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only