missing translation for 'onlineSavingsMsg'
Learn More

Zirconium fluoride, 99.5%, Thermo Scientific Chemicals

£63.21 - £182.91

Chemical Identifiers

CAS 7783-64-4
Molecular Formula F4Zr
Molecular Weight (g/mol) 167.21
MDL Number MFCD00011307
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10792761
View Documents
Thermo Scientific Acros
299790250
25 g Plastic bottle
£63.21
25g
Please to purchase this item. Need a web account? Register with us today!
10702771
View Documents
Thermo Scientific Acros
299791000
100 g Plastic bottle
£182.91
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7783-64-4
F4Zr
167.21
MFCD00011307

Specifications

905.0°C
7783-64-4
Fine Crystalline Powder
F4Zr
MFCD00011307
Solubility in water: 1.388 g/100mL. Other solubilities: slightly soluble in hf
167.21
Zirconium fluoride, 99.5%
640.0°C
White
99.4% min
ZrF4
15, 10375
167.21
99.5%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 232-018-1

missing translation for 'rtecsNumber' : ZH7875000

missing translation for 'tsca' : TSCA

Product Identifier
  • Zirconium fluoride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Zirconium fluoride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only