missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™, Thermo Scientific Chemicals

£28.74 - £148.74

Chemical Identifiers

CAS 23519-77-9, 71-23-8
Molecular Formula C12H28O4Zr
Molecular Weight (g/mol) 327.58
MDL Number MFCD00015307
InChI Key XPGAWFIWCWKDDL-UHFFFAOYSA-N
Synonym zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt
PubChem CID 90139
SMILES CCCO[Zr](OCCC)(OCCC)OCCC
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10356694
View Documents Promotion Details
Thermo Scientific Acros
427931000
100 mL AcroSeal™ Glass Bottle
£28.74
100mL
Please to purchase this item. Need a web account? Register with us today!
10694175
View Documents
Thermo Scientific Acros
427938000
800 mL AcroSeal™ Glass Bottle
£148.74
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

23519-77-9, 71-23-8
327.58
XPGAWFIWCWKDDL-UHFFFAOYSA-N
90139
C12H28O4Zr
MFCD00015307
zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt
CCCO[Zr](OCCC)(OCCC)OCCC

Specifications

23519-77-9, 71-23-8
1.0440g/mL
27.0 to 29% (ZrO2)
Zr(OCH2CH2CH3)4
1.044
XPGAWFIWCWKDDL-UHFFFAOYSA-N
zirconium(4+) tetrakis(propan-1-olate)
90139
ca. 70%
Zirconium(IV) propoxide, ca. 70%
Amber to Colorless
23°C
C12H28O4Zr
MFCD00015307
zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt
CCCO[Zr](OCCC)(OCCC)OCCC
327.58
327.57
Slightly Viscous Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 245-711-9

Product Identifier
  • Zirconium(IV) propoxide, solution in propanol

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only