missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium standard solution, for AAS, 1mg/mL Zr in 5% HF, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopySpecifications
Zirconium standard solution | |
1 mg/ml Zr in 5% HF | |
Plastic bottle | |
Colorless | |
MFCD00011303 MFCD00148878 | |
[Zr] | |
91.22 | |
CHEBI:33342 | |
Liquid |
990 to 1010μg/mL of Zr(4+) in 5% HF | |
7440-67-7, 7664-39-3, 7732-18-5 | |
100 mL | |
Zr | |
QCWXUUIWCKQGHC-UHFFFAOYSA-N | |
zirconium | |
23995 | |
AAS |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Zirconium standard solution, 1 mg/ml Zr in 5% HF
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H310-Fatal in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only