Learn More
Zirconium standard solution, for AAS, 1mg/mL Zr in 5% HF, Thermo Scientific Chemicals
£17.30
Specifications
Chemical Name or Material | Zirconium standard solution |
---|---|
Concentration | 990 to 1010μg/mL of Zr(4+) in 5% HF |
Name Note | 1 mg/ml Zr in 5% HF |
CAS | 7440-67-7, 7664-39-3, 7732-18-5 |
Color | Colorless |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopySpecifications
Zirconium standard solution | |
1 mg/ml Zr in 5% HF | |
Colorless | |
MFCD00011303 MFCD00148878 | |
[Zr] | |
91.22 | |
CHEBI:33342 | |
Liquid |
990 to 1010μg/mL of Zr(4+) in 5% HF | |
7440-67-7, 7664-39-3, 7732-18-5 | |
Zr | |
QCWXUUIWCKQGHC-UHFFFAOYSA-N | |
zirconium | |
23995 | |
AAS |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Zirconium standard solution, 1 mg/ml Zr in 5% HF
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H310-Fatal in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only