missing translation for 'onlineSavingsMsg'
Learn More

ZNF654 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Bio-Techne NBP3-10966-100UL

 View more versions of this product

Product Code. 18317245

  • £358.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ZNF654 Polyclonal specifically detects ZNF654 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZNF654
Polyclonal
Western Blot 1.0 ug/ml
FLJ10997, FLJ21142, zinc finger protein 654
The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF654 (NP_060763). Peptide sequence GRKFRNRGLMQKHLKNHVKKIQRQQIAAAQQDDQEVTALEEINCSSSSIS
100 μg
Primary
Human
Purified
Western Blot
Unconjugated
PBS buffer, 2% sucrose
Rabbit
Affinity purified
RUO
55279
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.