Learn More
ZNF804A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540123
Description
Recommended positive controls: U87-MG whole cell lysate or nuclear extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by ZNF804A is a zinc finger binding protein. Polymorphisms in this gene, especially rs1344706, are thought to confer increased susceptibility to schizophrenia, bipolar disorder, and heroin addiciton.
Specifications
ZNF804A | |
Polyclonal | |
Unconjugated | |
ZNF804A | |
C2orf10; zinc finger protein 804A; ZNF804A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91752 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
2.44 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q7Z570 | |
ZNF804A | |
Recombinant protein encompassing a sequence within the center region of human ZNF804A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.