missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 200 μL
Clear, autoclavable tips; color-coded racks.
£113.54 - £760.00
Specifications
Volume (Metric) | 200 μL |
---|---|
Filtered | Filtered |
Sterility | Sterile |
Color | Clear |
Rack Color | Blue |
Product Code | Brand | Tip Style | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tip Style | Description | Price | Quantity & Availability | ||||
12756769
|
Axygen™
ZTF-200-R-S |
Automation, Round | Standard |
£685.00
Pack of 4800 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||||
11384335
|
Axygen™
ZTF-200-R-S |
Automation, Round | Standard |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12746769
|
Axygen™
ZTF-200-L-R-S |
Automation, Round | Maxymum Recovery |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11374335
|
Axygen™
ZTF-200-L-R-S |
Automation, Round | Maxymum Recovery |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Clear Tips
- Format: 96 well
- Propylene construction
- Certified RNase/DNase free
- Nonpyrogenic
- Aerosol resistant filters
- Sterile
- Compatibility: Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head, Zymark™/Caliper RapidPlate™/SciClone™ with HV96 head/CAS-4200-1ch.-20-150μL/sec. head
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Specifications
200 μL | |
Sterile | |
Blue | |
Non-ANSI | |
Rack | |
Non-SBS Rack | |
Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head | |
Polyethylene |
Filtered | |
Clear | |
Autoclavable | |
Maxymum Recovery™ to further improve accuracy and precision while reducing sample loss | |
Automation, Round | |
50 Racks/Cs. | |
Caliper Zymark |