missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Zymosan, TLR2 Ligand

Applications: Functional, In vitro assay, In vivo assay, Ligand Activation

Brand:  Novus Biologicals™ NBP2-26233

Product Code. 18779873

  • £215.00 / 200mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Buffer: The Zymosan suspension solution is insoluble and remains opaque. Resuspend the solution by vortexing right before use. Dilution: Functional, In vitro assay, Ligand Activation Reconstitute with 50mg Zymosan with 1ml ethanol in a sterilized non-pyrogenic vial by vortexing to a final concentration of 50mg/ml. When diluting further, use sterilized endotoxin-free water or PBS
Specifications

Specifications

Store at -20°C in powder form. Store at -80°C once reconstituted.
This product is useful for activation of Toll-like Receptor 2 (TLR2). Stimulation of TLR2 has been reported with 1 to 10μL. Note: TLR2 activation at this range of concentrations of Zymosan has also been observed in our product validation tests. Use in In vivo assay and functional inhibition reported in scientific literature (PMID 25846753). Use in ligand activation reported in scientific literature (PMID 25957979). Use in in vitro reported in scientific literature (PMID 27351455).
Human
200 mg
Zymosan, TLR2 Ligand
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers