missing translation for 'onlineSavingsMsg'
Learn More
1,1,2,2-Tetrabromoethane, 98%, Thermo Scientific Chemicals Product Code.: 10751841

1,1,2,2-Tetrabromoethane, 98%, Thermo Scientific Chemicals

Product Code. 10751841
1 kg, Glass bottle
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10751841

Brand: Thermo Scientific Acros 180870010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-27-6
Molecular Formula C2H2Br4
Molecular Weight (g/mol) 345.65
MDL Number MFCD00000133
InChI Key QXSZNDIIPUOQMB-UHFFFAOYSA-N
Synonym acetylene tetrabromide, muthmann's liquid, ethane, 1,1,2,2-tetrabromo, tetrabromoacetylene, sym-tetrabromoethane, s-tetrabromoethane, 1,1,2,2-tetrabromaethan, 1,1,2,2-tetrabromoetano, tbe, unii-v00gkc584oShow MoreShow Less
PubChem CID 6588
IUPAC Name 1,1,2,2-tetrabromoethane
SMILES BrC(Br)C(Br)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 1.0°C
Density 2.9600g/mL
Boiling Point 244.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Br2CHCHBr2
Refractive Index 1.6355 to 1.6380
Quantity 1 kg
Beilstein 01, 94
Merck Index 15, 9330
Specific Gravity 2.96
Solubility Information Solubility in water: 0.63g/L (20°C). Other solubilities: soluble in acetone, benzene, miscible with ethanol, ether, chloroform, aniline, and glacial acetic acid
Formula Weight 345.64
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,1,2,2-Tetrabromoethane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0452203 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number180870010,180870025,180872500
Lot NumberA0408421 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number180870010,180870025,180872500
Lot NumberA0384954 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number180870010,180870025,180872500
Lot NumberA0447768 Certificate TypeCertificate of Analysis Date02/12/2022 Catalog Number180870010,180870025,180872500
Lot NumberA0446125 Certificate TypeCertificate of Analysis Date27/09/2022 Catalog Number180870010,180870025,180872500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,1,2,2-Tetrabromoethane, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.