missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(1,3-Benzodioxol-5-yl)-2-bromoethan-1-one, Thermo Scientific™
£33.40 - £283.00
Chemical Identifiers
CAS | 40288-65-1 |
---|---|
Molecular Formula | C9H7BrO3 |
Molecular Weight (g/mol) | 243.056 |
InChI Key | QBXCVQVFPVXAGS-UHFFFAOYSA-N |
PubChem CID | 243777 |
IUPAC Name | 1-(1,3-benzodioxol-5-yl)-2-bromoethanone |
SMILES | C1OC2=C(O1)C=C(C=C2)C(=O)CBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10722103
|
Thermo Scientific Maybridge
CC01605CB |
250 mg | Amber Glass Bottle |
£33.40
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10511692
|
Thermo Scientific Maybridge
CC01605DA |
1 g | Amber Glass Bottle |
£47.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10133214
|
Thermo Scientific Maybridge
CC01605EA |
10 g | Amber Glass Bottle |
£283.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
40288-65-1 | |
243.056 | |
243777 | |
C1OC2=C(O1)C=C(C=C2)C(=O)CBr |
C9H7BrO3 | |
QBXCVQVFPVXAGS-UHFFFAOYSA-N | |
1-(1,3-benzodioxol-5-yl)-2-bromoethanone |
Specifications
40288-65-1 | |
C9H7BrO3 | |
QBXCVQVFPVXAGS-UHFFFAOYSA-N | |
1-(1,3-benzodioxol-5-yl)-2-bromoethanone | |
243777 | |
97% |
97% | |
1-1,3-benzodioxol-5-yl-2-bromoethan-1-one, 1-1,3-benzodioxol-5-yl-2-bromoethanone, 1-2h-1,3-benzodioxol-5-yl-2-bromoethan-1-one, 5-bromoacetyl-1,3-benzodioxolane, ethanone,1-1,3-benzodioxol-5-yl-2-bromo, 1-benzo d 1,3 dioxol-5-yl-2-bromoethanone, 1-benzo 1,3 dioxol-5-yl-2-bromo-ethanone, 1-2h-1,3-benzodioxol-5-yl-2-bromoethanone, ethanone, 1-1,3-benzodioxol-5-yl-2-bromo, 1-2h-benzo 3,4-d 1,3-dioxolen-5-yl-2-bromoethan-1-one | |
C1OC2=C(O1)C=C(C=C2)C(=O)CBr | |
243.056 | |
243.06 | |
1-(1, 3-Benzodioxol-5-yl)-2-bromoethan-1-one |