missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,4,4-Tetramethyl-1,2,3,4-tetrahydronaphthaline, Technical Grade, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
100 g
Specifications
Specifications
Assay Percent Range | 90% |
Packaging | Amber glass bottle |
Quantity | 10 g |
Synonym | 1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene, 1,2,3,4-tetrahydro-1,1,4,4-tetramethylnaphthalene, 1,1,4,4-tetramethyltetralin, naphthalene, 1,2,3,4-tetrahydro-1,1,4,4-tetramethyl, naphthalene,1,2,3,4-tetrahydro-1,1,4,4-tetramethyl, 1,2,3,4-tetrahydro-1,1,4,4tetramethylnaphthalene, 1,1,4,4-tetramethyl-1,2,3,4-tetrahydro naphthalene, 1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthaline, 1,1,4,4-tetramethyl1,2,3,4-tetrahydro naphthalene, 1,2,3,4-tetrahydro-1,1,4,4-tetramethyl-naphthalene |
Formula Weight | 188.31 |
Percent Purity | 90% |
Chemical Name or Material | 1,1,4,4-Tetramethyl-1,2,3,4-tetrahydronaphthalene |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1,1,4,4-Tetramethyl-1,2,3,4-tetrahydronaphthaline, Technical Grade, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction