missing translation for 'onlineSavingsMsg'
Learn More
(+)-1,1'-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]ferrocene, 97+%, Thermo Scientific Chemicals Product Code.: 15405706

(+)-1,1'-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]ferrocene, 97+%, Thermo Scientific Chemicals

Product Code. 15405706
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15405706

Brand: Thermo Scientific Alfa Aesar H60364.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147762-89-8
Molecular Formula C26H40FeP2
Molecular Weight (g/mol) 470.40
MDL Number MFCD09265149
InChI Key MXOVWRYEUJWIFP-UHFFFAOYNA-N
Synonym +-1,1-bis 2r,5r-2,5-diethyl-1-phospholanyl ferrocene, 97+%
PubChem CID 131675863
SMILES [Fe].CC[C@@H]1CC[C@@H](CC)P1c1cccc1.CC[C@@H]1CC[C@@H](CC)P1c1cccc1

Specifications

Melting Point 85°C to 91°C
Quantity 1 g
Sensitivity Air Sensitive
Optical Rotation +380° (c=1 in Chloroform)
IUPAC Name carbanide;(2R,5R)-1-cyclopentyl-2,5-diethylphospholane;iron(2+)
Formula Weight 470.39
Percent Purity ≥97%
Chemical Name or Material (+)-1,1'-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]ferrocene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberK21Y035 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60364.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(+)-1,1'-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]ferrocene, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.