missing translation for 'onlineSavingsMsg'
Learn More
1,2,2,6,6-Pentamethyl-4-piperidinol, 99%, Thermo Scientific Chemicals Product Code.: 10058521

1,2,2,6,6-Pentamethyl-4-piperidinol, 99%, Thermo Scientific Chemicals

Product Code. 10058521
5 g, Glass bottle
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10058521

Brand: Thermo Scientific Acros 300860050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2403-89-6
Molecular Formula C10H21NO
Molecular Weight (g/mol) 171.28
MDL Number MFCD00191220
InChI Key NWHNXXMYEICZAT-UHFFFAOYSA-N
Synonym 1,2,2,6,6-pentamethyl-4-piperidinol, 4-piperidinol, 1,2,2,6,6-pentamethyl, 4-hydroxy-1,2,2,6,6-pentamethylpiperidine, unii-lr520bam5v, 1,2,2,6,6-pentamethyl-4-hydroxypiperidine, lr520bam5v, methyl-taa-ol, hopemp, pubchem22251, acmc-209g8aShow MoreShow Less
PubChem CID 75472
IUPAC Name 1,2,2,6,6-pentamethylpiperidin-4-ol
SMILES CC1(CC(CC(N1C)(C)C)O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 70°C to 76°C
Color Yellow
Density 0.9670g/mL
Boiling Point 238°C
Flash Point 118°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Specific Gravity 0.967
Solubility Information Solubility in water: in water: 4.8% (20°C). Other solubilities: in ethanol: 55% (20°C),in acetone: 38% (20°C),in chloroform: 52% (20°C)
Viscosity 15.3 mPa.s (80°C)
Formula Weight 171.28
Percent Purity 99%
Physical Form Flakes
Chemical Name or Material 1,2,2,6,6-Pentamethyl-4-piperidinol, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457527 Certificate TypeCertificate of Analysis Date22/11/2023 Catalog Number300862500,300860010,300860050
Lot NumberA0362390 Certificate TypeCertificate of Analysis Date13/01/2023 Catalog Number
Lot NumberA0324557 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number
Lot NumberA0358030 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2,2,6,6-Pentamethyl-4-piperidinol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.