missing translation for 'onlineSavingsMsg'
Learn More
1-(2-Bromoethyl)naphtalene, 97%, Thermo Scientific Chemicals Product Code.: 10716495

1-(2-Bromoethyl)naphtalene, 97%, Thermo Scientific Chemicals

Product Code. 10716495
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10716495

Brand: Thermo Scientific Acros 399560010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13686-49-2
Molecular Formula C12H11Br
Molecular Weight (g/mol) 235.12
MDL Number MFCD00037737
InChI Key GPHCPUFIWQJZOI-UHFFFAOYSA-N
Synonym 1-2-bromoethyl naphthalene, 2-1-naphthyl ethyl bromide, naphthalene, 1-2-bromoethyl, 2-bromoethylnaphthalene, 1-2-bromoethyl naphtalene, 1-2-bromo-ethyl-naphthalene, naphthalene,1-2-bromoethyl, 2-alpha-naphthyl ethyl bromide, naphthylathylbromid, 2-bromoethyl napthaleneShow MoreShow Less
PubChem CID 139541
IUPAC Name 1-(2-bromoethyl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2CCBr

Specifications

CAS Min % 97.0
CAS Max % 100.0
Density 1.6460g/mL
Boiling Point 237°C
Flash Point 110°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.646
Formula Weight 235.12
Percent Purity 97%
Chemical Name or Material 1-(2-Bromoethyl)naphtalene
Show More Show Less
compliance-icons
Product Identifier
  • 1-(2-Bromoethyl)naphtalene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(2-Bromoethyl)naphtalene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.