missing translation for 'onlineSavingsMsg'
Learn More
1,2-Epoxy-7-octene, 97%, Thermo Scientific Chemicals Product Code.: 11327106

1,2-Epoxy-7-octene, 97%, Thermo Scientific Chemicals

Product Code. 11327106
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11327106

Brand: Thermo Scientific Alfa Aesar L07033.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,2-Epoxy-7-octene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxy-7-octene is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19600-63-6
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00005156
InChI Key UKTHULMXFLCNAV-UHFFFAOYSA-N
Synonym 1,2-epoxy-7-octene, oxirane, 5-hexenyl, 5-hexenyloxirane, 7,8-epoxyoctene, ccris 3749, 2-hex-5-en-1-yl oxirane, oxirane, 5-hexenyl-, s, 2-hex-5-en-yloxirane, 2-5-hexenyl oxirane, acmc-20ap64Show MoreShow Less
PubChem CID 29678
IUPAC Name 2-hex-5-enyloxirane
SMILES C=CCCCCC1CO1

Specifications

Density 0.85
Boiling Point 167°C
Flash Point 38°C (100°F)
Refractive Index 1.435
Quantity 5 g
UN Number UN1993
Beilstein 1305290
Solubility Information Not miscible in water.
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material 1,2-Epoxy-7-octene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252698 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog NumberL07033.06,L07033.14
Lot Number10240477 Certificate TypeCertificate of Analysis Date05/05/2022 Catalog NumberL07033.06,L07033.14
Lot NumberG31U020 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL07033.06,L07033.14
Lot NumberH23S001 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL07033.06,L07033.14
Lot NumberR13H034 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL07033.06,L07033.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Epoxy-7-octene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.