missing translation for 'onlineSavingsMsg'
Learn More
1-(2-Phenoxyphenyl)ethanone, 97%, Thermo Scientific™ Product Code.: 10552154

1-(2-Phenoxyphenyl)ethanone, 97%, Thermo Scientific™

Product Code. 10552154
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 10552154

Brand: Thermo Scientific Maybridge CC49928DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 26388-13-6
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD08271961
InChI Key KPBCVVSDGJBODL-UHFFFAOYSA-N
Synonym 1-2-phenoxyphenyl ethanone, 2-acetyldiphenyl ether, 1-acetyl-2-phenoxybenzene, ethanone, 1-2-phenoxyphenyl
PubChem CID 10703750
IUPAC Name 1-(2-phenoxyphenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1OC1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 212.25
Percent Purity 97%
Chemical Name or Material 1-(2-Phenoxyphenyl)ethanone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(2-Phenoxyphenyl)ethanone, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.