missing translation for 'onlineSavingsMsg'
Learn More
1,3-Cyclohexanediol, cis + trans, 98%, Thermo Scientific Chemicals Product Code.: 11326796

1,3-Cyclohexanediol, cis + trans, 98%, Thermo Scientific Chemicals

Product Code. 11326796
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11326796

Brand: Thermo Scientific Alfa Aesar L04110.06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,3-Cyclohexanediol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Cyclohexanediol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-01-8
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00039458
InChI Key RLMGYIOTPQVQJR-UHFFFAOYSA-N
Synonym 1,3-cyclohexanediol, resorcitol, 1,3-dihydroxycyclohexane, 1,3-benzenediol, hexahydro, cis-1,3-cyclohexandiol, trans-1,3-cyclohexanediol, trans-1,3-cyclohexanol, 1,3-cyclohexanediol,c&t, acmc-209ll9, acmc-209po7Show MoreShow Less
PubChem CID 10433
IUPAC Name cyclohexane-1,3-diol
SMILES C1CC(CC(C1)O)O

Specifications

Melting Point ∼30°C
Boiling Point 246°C to 247°C
Quantity 5 g
Beilstein 2036654
Solubility Information Soluble in water.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material 1,3-Cyclohexanediol, Cis + Trans

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Cyclohexanediol, cis + trans, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.