missing translation for 'onlineSavingsMsg'
Learn More
(+/-)-1,4-Diazabicyclo[4.4.0]decane, 98+%, Thermo Scientific Chemicals Product Code.: 15498937

(+/-)-1,4-Diazabicyclo[4.4.0]decane, 98+%, Thermo Scientific Chemicals

Product Code. 15498937
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15498937

Brand: Thermo Scientific Alfa Aesar H52305.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4430-75-5
Molecular Formula C8H16N2
Molecular Weight (g/mol) 140.23
MDL Number MFCD01098257
InChI Key ONHPOXROAPYCGT-UHFFFAOYNA-N
Synonym octahydro-1h-pyrido 1,2-a pyrazine, octahydro-2h-pyrido 1,2-a pyrazine, octahydro-pyrido 1,2-a pyrazine, 1,4-diazabicyclo 4.4.0 decane, octahydro-1h-pyrido 1,2-a piperazine, chembl98511, octahydropyrido 1,2-a pyrazine, 2,3,4,6,7,8,9,9a-octahydro-1h-pyrido 1,2-a pyrazine, +/--1,4-diazabicyclo, octahydropyrido 1,2-a-pyrazineShow MoreShow Less
PubChem CID 2876391
IUPAC Name octahydro-1H-pyrido[1,2-a]pyrazine
SMILES C1CCN2CCNCC2C1

Specifications

Boiling Point 68°C to 72°C (3 mmHg)
Quantity 250 mg
UN Number UN3267
Formula Weight 140.23
Percent Purity ≥98%
Chemical Name or Material (+/-)-1,4-Diazabicyclo[4.4.0]decane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY21I021 Certificate TypeCertificate of Analysis Date25/01/2023 Catalog NumberH52305.03,H52305.MD
Lot NumberK02W069 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52305.03,H52305.MD
Lot NumberQ17G003 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52305.03,H52305.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(+/-)-1,4-Diazabicyclo[4.4.0]decane, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.