missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-dioxaspiro[4.5]dec-8-ylamine, Thermo Scientific™
£139.00 - £346.00
Chemical Identifiers
CAS | 97096-16-7 |
---|---|
Molecular Formula | C8H15NO2 |
Molecular Weight (g/mol) | 157.21 |
MDL Number | MFCD04114556 |
InChI Key | KDAFVGCPLFJMHY-UHFFFAOYSA-N |
Synonym | 1,4-dioxaspiro 4.5 decan-8-amine, 1,4-dioxa-spiro 4.5 dec-8-ylamine, 8-amino-1,4-dioxaspiro 4,5 decane, 1,4-dioxaspiro 4.5 dec-8-ylamine, 1,4-dioxaspiro 4.5 decane-8-amine, 1,4-dioxa-spiro 4.5 dec-8-yl amine, 1,4-dioxa-spiro 4.5 dec-8-yl-amine |
PubChem CID | 14634315 |
IUPAC Name | 1,4-dioxaspiro[4.5]decan-8-amine |
SMILES | NC1CCC2(CC1)OCCO2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10506144
|
Thermo Scientific Maybridge
MO08523CB |
250 mg | Amber Glass Bottle |
N/A
|
N/A | ||||
10235854
|
Thermo Scientific Maybridge
MO08523DA |
1 g | Amber Glass Bottle |
N/A
|
N/A | ||||
Chemical Identifiers
97096-16-7 | |
157.21 | |
KDAFVGCPLFJMHY-UHFFFAOYSA-N | |
14634315 | |
NC1CCC2(CC1)OCCO2 |
C8H15NO2 | |
MFCD04114556 | |
1,4-dioxaspiro 4.5 decan-8-amine, 1,4-dioxa-spiro 4.5 dec-8-ylamine, 8-amino-1,4-dioxaspiro 4,5 decane, 1,4-dioxaspiro 4.5 dec-8-ylamine, 1,4-dioxaspiro 4.5 decane-8-amine, 1,4-dioxa-spiro 4.5 dec-8-yl amine, 1,4-dioxa-spiro 4.5 dec-8-yl-amine | |
1,4-dioxaspiro[4.5]decan-8-amine |
Specifications
97096-16-7 | |
MFCD04114556 | |
KDAFVGCPLFJMHY-UHFFFAOYSA-N | |
1,4-dioxaspiro[4.5]decan-8-amine | |
14634315 | |
97% |
C8H15NO2 | |
1,4-dioxaspiro 4.5 decan-8-amine, 1,4-dioxa-spiro 4.5 dec-8-ylamine, 8-amino-1,4-dioxaspiro 4,5 decane, 1,4-dioxaspiro 4.5 dec-8-ylamine, 1,4-dioxaspiro 4.5 decane-8-amine, 1,4-dioxa-spiro 4.5 dec-8-yl amine, 1,4-dioxa-spiro 4.5 dec-8-yl-amine | |
NC1CCC2(CC1)OCCO2 | |
157.21 | |
157.21 | |
1, 4-dioxaspiro[4.5]dec-8-ylamine |
Safety and Handling
GHS Signal Word: Danger
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1,4-dioxaspiro[4.5]dec-8-ylamine, Thermo Scientific™