missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Methylphenyl)-1H-pyrrole-2-carbaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS | 30186-38-0 |
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C12H11NO |
MDL Number | MFCD01567103 |
Quantity | 1 g |
Synonym | 1-4-methylphenyl-1h-pyrrole-2-carbaldehyde, 1-4-methylphenyl pyrrole-2-carbaldehyde, 1-p-tolyl-1h-pyrrole-2-carbaldehyde, 1-p-tolyl-1h-pyrrole-2-carboxaldehyde, 1-p-tolyl-2-formylpyrrole, 1h-pyrrole-2-carboxaldehyde, 1-4-methylphenyl, acmc-20apac, 1-4-methylphenyl pyrrole-2-carboxaldehyde |
InChI Key | LASYAEOJPVQNFO-UHFFFAOYSA-N |
SMILES | CC1=CC=C(C=C1)N1C=CC=C1C=O |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1-(4-Methylphenyl)-1H-pyrrole-2-carbaldehyde, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction