missing translation for 'onlineSavingsMsg'
Learn More
1-Bromo-4-phenylbutane, 98%, Thermo Scientific Chemicals Product Code.: 11490758

1-Bromo-4-phenylbutane, 98%, Thermo Scientific Chemicals

Product Code. 11490758
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11490758

Brand: Thermo Scientific Alfa Aesar B25237.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Bromo-4-phenylbutane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-phenylbutane is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13633-25-5
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.12
MDL Number MFCD00154988
InChI Key XPBQQAHIVODAIC-UHFFFAOYSA-N
Synonym 1-bromo-4-phenylbutane, 4-bromobutyl benzene, 4-phenylbutyl bromide, 4-phenyl 1-butyl bromide, 4-phenyl-1-butyl bromide, 4-phenyl-1-bromobutane, 1-bromo-4-phenyl-butane, benzene, 4-bromobutyl, benzene, bromobutyl, 4-phenyl-1-bromo butaneShow MoreShow Less
PubChem CID 259668
SMILES BrCCCCC1=CC=CC=C1

Specifications

Density 1.214
Boiling Point 131°C to 133°C (12 mmHg)
Quantity 1 g
Solubility Information Slightly soluble in water.
IUPAC Name (4-bromobutyl)benzene
Formula Weight 213.12
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-phenylbutane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromo-4-phenylbutane, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.