Learn More
1-Bromododecane, 98%, Thermo Scientific Chemicals
Descripción
1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Especificaciones
Especificaciones
Melting Point | -10°C |
Density | 1.04 |
Boiling Point | 276°C |
Flash Point | 146°C (294°F) |
Odor | Weak |
Linear Formula | CH3(CH2)11Br |
Refractive Index | 1.458 |
Quantity | 250 g |
Beilstein | 506159 |
Merck Index | 14,5389 |
Mostrar más |
Sugerencias de productos
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10251994 | Certificate of Analysis | 08/08/2024 | A10600.30,A10600.0I,A10600.0B |
10242407 | Certificate of Analysis | 29/12/2023 | A10600.30,A10600.0I,A10600.0B |
10248053 | Certificate of Analysis | 25/09/2023 | A10600.30,A10600.0I,A10600.0B |
10212804 | Certificate of Analysis | 07/08/2023 | A10600.30,A10600.0I,A10600.0B |
10224821 | Certificate of Analysis | 14/06/2022 | A10600.30,A10600.0I,A10600.0B |
Seguridad y manipulación

- 1-Bromododecane
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Lauryl bromide
RUO – Research Use Only
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.