missing translation for 'onlineSavingsMsg'
Learn More

1-Bromonaphthalene, 97%, Thermo Scientific Chemicals

£22.61 - £132.66

Chemical Identifiers

CAS 90-11-9
Molecular Formula C10H7Br
Molecular Weight (g/mol) 207.07
MDL Number MFCD00003868
InChI Key DLKQHBOKULLWDQ-UHFFFAOYSA-N
Synonym naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthalene
PubChem CID 7001
IUPAC Name 1-bromonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11498086
View Documents
Thermo Scientific Alfa Aesar
A14122.18
50 g
£22.61
50g
Please to purchase this item. Need a web account? Register with us today!
11408096
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14122.30
250 g
£46.83
250g
Please to purchase this item. Need a web account? Register with us today!
11488086
View Documents
Thermo Scientific Alfa Aesar
A14122.0B
1000 g
£132.66
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Bromonaphthalene is used in organic synthesis and refractometry of fats. It is used in the preparation of N-aryl imidazoles and diaryl ethers. It is used to determine the refractive index of crystals and the water content of alcohols. It is utilized in the preparation of glutathione peroxidase-like antioxidant activity of diaryl diselenides. Further, it serves as a solvent for the exfoliation and dispersion of hexabenzocoronene. In addition to this, it is used as a refrigerant and a solvent molecular weight substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromonaphthalene is used in organic synthesis and refractometry of fats. It is used in the preparation of N-aryl imidazoles and diaryl ethers. It is used to determine the refractive index of crystals and the water content of alcohols. It is utilized in the preparation of glutathione peroxidase-like antioxidant activity of diaryl diselenides. Further, it serves as a solvent for the exfoliation and dispersion of hexabenzocoronene. In addition to this, it is used as a refrigerant and a solvent molecular weight substance.

Solubility
Miscible with chloroform, ethanol, ether, benzene and ethyl acetate. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

90-11-9
207.07
DLKQHBOKULLWDQ-UHFFFAOYSA-N
7001
C1=CC=C2C(=C1)C=CC=C2Br
C10H7Br
MFCD00003868
naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthalene
1-bromonaphthalene

Specifications

90-11-9
1.485
>110°C (230°F)
C10H7Br
MFCD00003868
14,1425
Miscible with chloroform,ethanol,ether,benzene and ethyl acetate. Slightly miscible with water.
C1=CC=C2C(=C1)C=CC=C2Br
207.07
207.08
Liquid
°C to 2°C to 0°C
279°C to 281°C
97%
1.657
1906414
naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthalene
DLKQHBOKULLWDQ-UHFFFAOYSA-N
1-bromonaphthalene
7001
97%
1-Bromonaphthalene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H319

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 201-965-2

missing translation for 'rtecsNumber' : QJ1545000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • 1-Bromonaphthalene

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : 1-Bromonaphthalene

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only