missing translation for 'onlineSavingsMsg'
Learn More
1-Butanol, 99.5%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals Product Code.: 10321311

1-Butanol, 99.5%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals

Product Code. 10321311
1 L, EcoSafPak™ Glass Bottle
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10321311

Brand: Thermo Scientific Acros 423490010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropaneShow MoreShow Less
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

CAS Min % 99.4
CAS Max % 100.0
Density 0.8100g/mL
Quantity 1 L
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Assay Percent Range 99.5%
Residue after Evaporation 0.005% max.
Titratable Acid 0.0008 meq/g max.
Linear Formula CH3(CH2)3OH
Vapor Pressure Vapor Pressure: 6.7mbar at 20°C
Beilstein 01, 367
Water 0.1% max. (K.F.)
Merck Index 15, 1542
Formula Weight 74.12
Percent Purity ≥99.4%
Grade ACS Reagent
Chemical Name or Material 1-Butanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467255 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number423495000,423490025,423490010,423490100
Lot NumberA0468649 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number423495000,423490025,423490010,423490100
Lot NumberA0456530 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog Number423495000,423490025,423490010,423490100
Lot NumberA0453222 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number423495000,423490025,423490010,423490100
Lot NumberA0421535 Certificate TypeCertificate of Analysis Date04/04/2023 Catalog Number423495000,423490025,423490010,423490100
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Butanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Butanol, 99.5%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.