missing translation for 'onlineSavingsMsg'
Learn More
1-Butanol, ACS, 99.4+%, Thermo Scientific Chemicals Product Code.: 11347388

1-Butanol, ACS, 99.4+%, Thermo Scientific Chemicals

Product Code. 11347388
1 L
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
4 x 4 L
This item is not returnable. View return policy

Product Code. 11347388

Brand: Thermo Scientific Alfa Aesar 031068.K2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.005%, Titratable acid 0.0008meq/g, Carbonyl compounds (as butyraldehyde) 0.01%, Butyl ether 0.2%, H2O 0.1%

1-Butanol is used as solvent for many organic reactions in pharmaceutical and chemical industry, as a solvent in coating industries and also used as a diluent/reactant in the manufacture of urea-formaldehyde and melamine-formaldehyde resins. It is used in the production of drugs, alkaloids, additive in floor cleaners, consumer products and stain removers. It is used as mobile phase in paper and thin layer chromatography. It is also used as an alternative for gasoline and diesel fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropaneShow MoreShow Less
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Spécification

Color Colorless
Density 0.81
Boiling Point 117°C to 118°C
Quantity 1 L
Flash Point 35°C (95°F)
Physical Form Liquid
Refractive Index 1.399
Assay Percent Range 99.4+%
Beilstein 969148
Solubility Information Miscible with alcohols,ketones,aldehydes,ethers,aliphatic and aromatic hydrocarbons. Slightly miscible with water.
Formula Weight 74.12
Grade ACS
Concentration or Composition (by Analyte or Components) 1-Butanol
Chemical Name or Material 1-Butanol
Afficher plus Afficher moins

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW11K041 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog Number031068.M1,031068.AP,031068.K7,031068.K2,031068.M6
Lot NumberW10K048 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog Number031068.M1,031068.AP,031068.K7,031068.K2,031068.M6
Lot NumberM29K027 Certificate TypeCertificate of Analysis Date02/02/2024 Catalog Number031068.M1,031068.AP,031068.K7,031068.K2,031068.M6
Lot NumberX12J020 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog Number031068.M1,031068.AP,031068.K7,031068.K2,031068.M6
Lot NumberS22J025 Certificate TypeCertificate of Analysis Date28/06/2023 Catalog Number031068.M1,031068.AP,031068,031068.K7,031068.K2,031068.M6
compliance-icons compliance-icons compliance-icons
Identifiant produit
  • 1-Butanol
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informations supplémentaires
  • MIXTURE LIST-Contains : 1-Butanol

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit
1-Butanol, ACS, 99.4+%, Thermo Scientific Chemicals >

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis