missing translation for 'onlineSavingsMsg'
Learn More
1-Butylamine, 99%, Thermo Scientific Chemicals Product Code.: 11392216

1-Butylamine, 99%, Thermo Scientific Chemicals

Product Code. 11392216
500 mL
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11392216

Brand: Thermo Scientific Alfa Aesar L03575.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

1-Butylamine is used as an intermediate for pharmaceuticals, synthetic tanning agents, dyestuffs and emulsifiers. It also plays an important role in the preparation of N,N'-dibutylthiourea, a rubber vulcanization accelerator. It is involved in the preparation of n-butylbenzenesulfonamide, which is a plasticizer of nylon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Butylamine is used as an intermediate for pharmaceuticals, synthetic tanning agents, dyestuffs and emulsifiers. It also plays an important role in the preparation of N,N′-dibutylthiourea, a rubber vulcanization accelerator. It is involved in the preparation of n-butylbenzenesulfonamide, which is a plasticizer of nylon.

Solubility
Miscible with ethanol and water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, aluminum, copper, copper alloys and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-73-9
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00011690
InChI Key HQABUPZFAYXKJW-UHFFFAOYSA-N
Synonym butylamine, n-butylamine, 1-butanamine, 1-aminobutane, 1-butylamine, monobutylamine, n-butylamin, mono-n-butylamine, norvalamine, 1-aminobutanShow MoreShow Less
PubChem CID 8007
ChEBI CHEBI:43799
IUPAC Name butan-1-amine
SMILES CCCCN

Specifications

Melting Point -50°C
Density 0.738
Boiling Point 76°C to 78°C
Flash Point −14°C (6°F)
Odor Ammonia-like
Linear Formula CH3CH2CH2CH2NH2
Refractive Index 1.401
Quantity 500 mL
UN Number UN1125
Beilstein 605269
Sensitivity Air Sensitive
Merck Index 14,1543
Solubility Information Miscible with ethanol and water.
Formula Weight 73.14
Percent Purity 99%
Chemical Name or Material 1-Butylamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10240466 Certificate TypeCertificate of Analysis Date13/02/2024 Catalog NumberL03575.0F,L03575.AE,L03575.AP
Lot NumberA0455230 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberL03575.0F,L03575.AE,L03575.AP
Lot Number10239550 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL03575.0F,L03575.AE,L03575.AP
Lot Number10202729 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL03575.0F,L03575.AE,L03575.AP
Lot NumberM30A051 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL03575.0F,L03575.AE,L03575.AP
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Butylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : n-Butylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Butylamine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.