Learn More
Thermo Scientific™ Cartridges for Multidrop™ Pico 1 & Pico 8 Digital Dispensers
GREENER_CHOICE
The Dispensehead cartidges allow dispensing of one to eight fluids per cassette, from 11 picoliters to 10 microliters of small molecules and biomolecules in DMSO or aqueous solution
£108.00 - £1419.60
Specifications
Product Type Digital Dispenser Cartridge
Autoclavable Not Autoclavable
For Use With (Application) Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting
Interface USB
Plate Type 12, 24, 48, 96, 384, 1536 well plate, Plate height: 6-47 mm
View More Specs
Products 6
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
17961961
GREENER_CHOICE
View Documents
Thermo Scientific™
LTR0001
1-ch 20μL Cartridges for Multidrop Pico 1
£1320.00
Pack of 30
Estimated Shipment: 18-06-2025
to see stock.
Add to basket
17971961
GREENER_CHOICE
View Documents
Thermo Scientific™
LTR0002
1-ch 200μL Cartridges for Multidrop Pico 1
£1395.00
Pack of 30
Estimated Shipment: 18-06-2025
to see stock.
Add to basket
17991961
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
LTR0003
4-ch 200 μL Cartridges for Multidrop Pico 8
£1325.00
Pack of 20
In Stock
to see stock.
Add to basket
16643853
View Documents Promotion Details
Thermo Scientific™
LTR0003
4-ch 200 μL Cartridges for Multidrop Pico 8
£108.00
Each
In Stock
to see stock.
Edge
Add to basket
16653853
View Documents Promotion Details
Thermo Scientific™
LTR0004
8-ch 20 μL Cartridges for Multidrop Pico 8
£108.00
Each
In Stock
to see stock.
Edge
Add to basket
17981961
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
LTR0004
8-ch 20 μL Cartridges for Multidrop Pico 8
£1419.60
Pack of 20
In Stock
to see stock.
Edge
Add to basket
Description
Description
Great choice for dispensing picoliter to microliter volumes for one to eight different types of fluid across an entire plate or just part of a plate.
  • Cost Saving through assay miniaturization
  • Improved efficiency and sample integrity
  • Powerful, flexible, and easy to use software
Specifications
Specifications
Digital Dispenser Cartridge
Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting
12, 24, 48, 96, 384, 1536 well plate, Plate height: 6-47 mm
11 pL to 10 μL in seconds
100/240 V
Not Autoclavable
USB
≤ 8%
100 to 240V 50/60Hz
50/60 Hz
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Cartridges for Multidrop™ Pico 1 & Pico 8 Digital Dispensers