Learn More
Thermo Scientific™ Cartridges for Multidrop™ Pico 1 & Pico 8 Digital Dispensers
GREENER_CHOICE
The Dispensehead cartidges allow dispensing of one to eight fluids per cassette, from 11 picoliters to 10 microliters of small molecules and biomolecules in DMSO or aqueous solution
Brand:  Thermo Scientific™ LTR0004
1250.72 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
 View more versions of this product
Product Code. 17981961
  • £1419.60 / Pack of 20
In Stock
Edge
Explore more special offers
Add to basket
Description
Description
Great choice for dispensing picoliter to microliter volumes for one to eight different types of fluid across an entire plate or just part of a plate.
  • Cost Saving through assay miniaturization
  • Improved efficiency and sample integrity
  • Powerful, flexible, and easy to use software
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Digital Dispenser Cartridge
8-ch 20 μL Cartridges for Multidrop Pico 8
11 pL to 20 μL (8-well Cartridge), 1 nL to 200 μL (4-well Cartridge)
Mulitdrop Pico 8 Digital Dispenser
12, 24, 48, 96, 384, 1536 well plate, Plate height: 6-47 mm
PicoIT 8
8 x 10.5 x 2.5 cm
100 to 240V 50/60Hz
50/60 Hz
Not Autoclavable
8
Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting
USB
≤ 8%
11 pL to 10 μL in seconds
0.45
100/240 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Cartridges for Multidrop™ Pico 1 & Pico 8 Digital Dispensers > Thermo Scientific 8-ch 20μL Cartridge for Multidrop™ Pico 8 Digital Dispenser, dispense from 11pL up to 20μL