missing translation for 'onlineSavingsMsg'
Learn More
1-Dodecanesulfonyl chloride, 97%, Thermo Scientific Chemicals Product Code.: 11354514

1-Dodecanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11354514
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11354514

Brand: Thermo Scientific Alfa Aesar L01224.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10147-40-7
Molecular Formula C12H25ClO2S
Molecular Weight (g/mol) 268.84
MDL Number MFCD00014728
InChI Key PBULHKIPTBIZHO-UHFFFAOYSA-N
Synonym 1-dodecanesulfonyl chloride, 1-dodecanesulfonylchloride, unii-g45gs200po, 1-dodecanesulphonyl chloride, dodecylsulfonyl chloride, dodecane-1-sulphonyl chloride, chlorododecylsulfone, dodecanesulfonylchloride, acmc-20a5qf, dodecanesulfonyl chlorideShow MoreShow Less
PubChem CID 66280
IUPAC Name dodecane-1-sulfonyl chloride
SMILES CCCCCCCCCCCCS(Cl)(=O)=O

Specifications

Melting Point 39°C to 42°C
Boiling Point 126°C to 130°C (4 mmHg)
Linear Formula CH3(CH2)11SO2Cl
Quantity 5 g
UN Number UN3261
Beilstein 1778092
Sensitivity Moisture sensitive
Formula Weight 268.84
Percent Purity ≥98%
Chemical Name or Material 1-Dodecanesulfonyl chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10232814 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog NumberL01224.06,L01224.14
Lot Number10242882 Certificate TypeCertificate of Analysis Date16/08/2022 Catalog NumberL01224.06,L01224.14
Lot NumberFA000496 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL01224.06,L01224.14
Lot Number10232814 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL01224.06,L01224.14
Lot Number10230688 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL01224.06,L01224.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Dodecanesulfonyl chloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.