missing translation for 'onlineSavingsMsg'
Learn More
1-Iodooctane, 98+%, stab. with copper, Thermo Scientific Chemicals Product Code.: 11405846

1-Iodooctane, 98+%, stab. with copper, Thermo Scientific Chemicals

Product Code. 11405846
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11405846

Brand: Thermo Scientific Alfa Aesar B22214.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alkylation of [2,2?-([2,2?-bipyridine]-6,6?-diyl)bis[phenolato]-N,N?,O,O?]nickel(II)was performed using the catalytic reduction of 1-iodooctane. Primary alkyl iodide 1-iodooctane reacts with lithium aluminum deuteride (LAD) in a 1:0.2 ratio to form 1-deuteriooctane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alkylation of [2,2′-([2,2′-bipyridine]-6,6′-diyl)bis[phenolato]-N,N′,O,O′]nickel(II)was performed using the catalytic reduction of 1-iodooctane. Primary alkyl iodide 1-iodooctane reacts with lithium aluminum deuteride (LAD) in a 1:0.2 ratio to form 1-deuteriooctane.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-27-6
Molecular Formula C8H17I
Molecular Weight (g/mol) 240.128
MDL Number MFCD00001105
InChI Key UWLHSHAHTBJTBA-UHFFFAOYSA-N
Synonym n-octyl iodide, octyl iodide, octane, 1-iodo, 1-octyl iodide, 1-n-octyl iodide, 1-jodoktan, 1-jodoktan czech, 1-iodooctane;n-octyl iodide, iodoctane, iodooctaneShow MoreShow Less
PubChem CID 12380
IUPAC Name 1-iodooctane
SMILES CCCCCCCCI

Specifications

Melting Point -46°C
Density 1.325
Boiling Point 225°C to 226°C
Flash Point 95°C (203°F)
Linear Formula CH3(CH2)7I
Refractive Index 1.4878
Quantity 25 g
Beilstein 1697479
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 240.13
Percent Purity ≥98%
Chemical Name or Material 1-Iodooctane, stabilized with copper
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241044 Certificate TypeCertificate of Analysis Date10/06/2022 Catalog NumberB22214.14,B22214.36,B22214.22
Lot Number10240114 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberB22214.14,B22214.36,B22214.22
Lot Number10176825 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22214.14,B22214.36,B22214.22
Lot Number10200992 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22214.14,B22214.36,B22214.22
Lot Number10209225 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22214.14,B22214.36,B22214.22
compliance-icons
Product Identifier
  • 1-Iodooctane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1-Iodooctane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Iodooctane, 98+%, stab. with copper, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.