missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-2-pyrrolidinone, ACS Reagent, Honeywell
£41.61 - £196.31
Chemical Identifiers
CAS | 872-50-4 |
---|---|
Molecular Formula | C5H9NO |
Molecular Weight (g/mol) | 99.133 |
MDL Number | MFCD00003193 |
InChI Key | SECXISVLQFMRJM-UHFFFAOYSA-N |
Synonym | 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl |
PubChem CID | 13387 |
ChEBI | CHEBI:7307 |
IUPAC Name | 1-methylpyrrolidin-2-one |
SMILES | CN1CCCC1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15695540
|
Honeywell Riedel-de Haën
443778-500ML |
500 mL | N/A | ||||||
15665540
|
Honeywell Riedel-de Haën
443778-1L |
1 L |
£92.05
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15675540
|
Honeywell Riedel-de Haën
443778-2.5L |
2.5 L |
£196.31
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
872-50-4 | |
99.133 | |
SECXISVLQFMRJM-UHFFFAOYSA-N | |
13387 | |
1-methylpyrrolidin-2-one |
C5H9NO | |
MFCD00003193 | |
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl | |
CHEBI:7307 | |
CN1CCCC1=O |
Specifications
872-50-4 | |
0.095 | |
518°F | |
MFCD00003193 | |
106420 | |
SECXISVLQFMRJM-UHFFFAOYSA-N | |
1-methylpyrrolidin-2-one | |
13387 | |
99.13g/mol | |
ACS Reagent |
1.028g/mL at 25°C | |
91°C (195.8°F) | |
C5H9NO | |
0.29 mmHg (20°C), 0.99 mmHg (40°C) | |
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl | |
CN1CCCC1=O | |
99.133 | |
CHEBI:7307 | |
≥99% | |
1-Methyl-2-pyrrolidinone |
Safety and Handling
P201-P280-P305 + P351 + P338-P308 + P313
Danger
missing translation for 'einecsNumber' : 212-828-1