missing translation for 'onlineSavingsMsg'
Learn More
1-Methylimidazole, 99%, Thermo Scientific Chemicals Product Code.: 11404284

1-Methylimidazole, 99%, Thermo Scientific Chemicals

Product Code. 11404284
100 g
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11404284

Brand: Thermo Scientific Alfa Aesar A12575.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Methylimidazole is used as a precursor for the synthesis of pyrrole-imidazole polyamides, ionic liquids such as 1-butyl-3-methylimidazolium hexafluorophosphate. It is actively involved in removing acid during the production of diethoxyphenylphosphine. It is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylimidazole is used as a precursor for the synthesis of pyrrole-imidazole polyamides, ionic liquids such as 1-butyl-3-methylimidazolium hexafluorophosphate. It is actively involved in removing acid during the production of diethoxyphenylphosphine. It is used as an intermediate in organic synthesis.

Solubility
Miscible with water.

Notes
Incompatible with carbon dioxide and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-47-7
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005292
InChI Key MCTWTZJPVLRJOU-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazoleShow MoreShow Less
PubChem CID 1390
ChEBI CHEBI:113454
SMILES CN1C=CN=C1

Specifications

Melting Point -6°C
Density 1.031
Boiling Point 195°C to 197°C
Flash Point 92°C (197°F)
Refractive Index 1.497
Quantity 100 g
UN Number UN3267
Beilstein 105197
Sensitivity Hygroscopic
Solubility Information Miscible with water.
IUPAC Name 1-methyl-1H-imidazole
Formula Weight 82.11
Percent Purity 99%
Chemical Name or Material 1-Methylimidazole
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253120 Certificate TypeCertificate of Analysis Date18/11/2024 Catalog NumberA12575.36,A12575.A3,A12575.22
Lot Number10252278 Certificate TypeCertificate of Analysis Date02/09/2024 Catalog NumberA12575.36,A12575.A3,A12575.22
Lot Number10247820 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog NumberA12575.36,A12575.A3,A12575.22
Lot Number5001I23T Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA12575.36,A12575.A3,A12575.22
Lot Number5001I27N Certificate TypeCertificate of Analysis Date02/05/2023 Catalog NumberA12575.36,A12575.A3,A12575.22
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Methyl-1H-imidazol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1-Methyl-1H-imidazol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methylimidazole, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.