missing translation for 'onlineSavingsMsg'
Learn More
1-Naphthoyl chloride, 99%, Thermo Scientific Chemicals Product Code.: 10559263

1-Naphthoyl chloride, 99%, Thermo Scientific Chemicals

Product Code. 10559263
50 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10559263

Brand: Thermo Scientific Acros 270380500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 879-18-5
Molecular Formula C11H7ClO
Molecular Weight (g/mol) 190.63
MDL Number MFCD00004002
InChI Key NSNPSJGHTQIXDO-UHFFFAOYSA-N
Synonym 1-naphthoyl chloride, 1-naphthalenecarbonyl chloride, 1-naphthoylchloride, 1-naphthoic acid chloride, 1-chlorocarbonyl naphthalene, alpha-naphthoyl chloride, .alpha.-naphthoyl chloride, naphthalenecarbonyl chloride, naphthoylchloride, naphthoyl chlorideShow MoreShow Less
PubChem CID 70146
IUPAC Name naphthalene-1-carbonyl chloride
SMILES ClC(=O)C1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 16°C to 19°C
Color White
Density 1.2600g/mL
Boiling Point 190°C (35.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6515 to 1.6535
Quantity 50 g
Beilstein 09, 648
Specific Gravity 1.26
Solubility Information Solubility in water: decomposes
Formula Weight 190.63
Percent Purity 99%
Physical Form Crystals, Powder or Flakes
Chemical Name or Material 1-Naphthoyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0416902 Certificate TypeCertificate of Analysis Date13/01/2023 Catalog Number270380500,270380100
Lot NumberA0394938 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number270380500,270380100
Lot NumberA0412274 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number270380500,270380100
Lot NumberA0410276 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number270380500,270380100
Lot NumberA0416902 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number270380500,270380100
compliance-icons
Product Identifier
  • 1-Naphthoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Naphthoyl chloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.