missing translation for 'onlineSavingsMsg'
Learn More
1-Octanol, 99%, Thermo Scientific Chemicals Product Code.: 11428556

1-Octanol, 99%, Thermo Scientific Chemicals

Product Code. 11428556
2500 mL
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
This item is not returnable. View return policy

Product Code. 11428556

Brand: Thermo Scientific Alfa Aesar A15977.0F

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.

Solubility
Miscible with ethanol, ether and propylene glycol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcoholShow MoreShow Less
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO

Specifications

Melting Point -16°C
Density 0.825
Boiling Point 193°C to 195°C
Flash Point 81°C (177°F)
Assay Percent Range 99%
Odor Sweet
Linear Formula CH3(CH2)7OH
Refractive Index 1.429
Quantity 2500 mL
Beilstein 1697461
Merck Index 14,6751
Solubility Information Miscible with ethanol,ether and propylene glycol. Immiscible with water.
Formula Weight 130.23
Percent Purity 99%
Chemical Name or Material 1-Octanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462306 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA15977.AP,A15977.0D,A15977.0F
Lot NumberA0455836 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog NumberA15977.AP,A15977.0D,A15977.0F
Lot NumberA0457508 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog NumberA15977.AP,A15977.0D,A15977.0F
Lot NumberA0453932 Certificate TypeCertificate of Analysis Date10/08/2023 Catalog NumberA15977.AP,A15977.0D,A15977.0F
Lot NumberA0454684 Certificate TypeCertificate of Analysis Date10/08/2023 Catalog NumberA15977.AP,A15977.0D,A15977.0F
compliance-icons
Product Identifier
  • 1-Octanol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-octanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Octanol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.