Learn More
1-Propanol, for HPLC, Thermo Scientific Chemicals
£36.90 - £78.90
Chemical Identifiers
CAS | 71-23-8 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.10 |
MDL Number | MFCD00002941 |
InChI Key | BDERNNFJNOPAEC-UHFFFAOYSA-N |
Synonym | 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol |
PubChem CID | 1031 |
ChEBI | CHEBI:28831 |
IUPAC Name | propan-1-ol |
SMILES | CCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10455465
|
Thermo Scientific Acros
|
1 L | Glass bottle |
£36.90
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10791464
|
Thermo Scientific Acros
|
2.5 L | Glass bottle |
£78.90
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLCChemical Identifiers
71-23-8 | |
60.10 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
1031 | |
propan-1-ol |
C3H8O | |
MFCD00002941 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
CHEBI:28831 | |
CCCO |
Specifications
-127.0°C | |
71-23-8 | |
0.8000g/mL | |
15°C | |
1.3840 to 1.3860 | |
C3H8O | |
MFCD00002941 | |
15, 7955 | |
Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents | |
CCCO | |
60.10 | |
CHEBI:28831 | |
≥99.8% | |
1-Propanol |
0.05 max. (1 cm cell vs HPLC−grade water) at 250nm, 0.3 max. (1 cm cell vs HPLC−grade water) at 220nm, 0.7 max. (1 cm cell vs HPLC−grade water) at 210nm | |
Colorless | |
97.0°C | |
Liquid | |
0.8 | |
CH3CH2CH2OH | |
2.2 mPa.s (20°C) | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
propan-1-ol | |
1031 | |
60.1 | |
HPLC |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Causes serious eye damage.
Highly flammable liquid and vapor.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-746-9
- 1-Propanol
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only